COM448685531 Combi-Blocks, Inc.

3-(4-Chlorophenyl)-5-trifluoromethylbenzoic acid

  • MDL number MFCD18319938
  • Cas Number : 1261964-03-7
  • Molecular Weight : 300.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0067

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0067
Availability details for
Quantity
  • Availability
  • Add to Cart