COM448685532 Combi-Blocks, Inc.

4-(2,4-Difluorophenyl)-2-fluorobenzoic acid

  • MDL number MFCD12859432
  • Cas Number : 505082-86-0
  • Molecular Weight : 252.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0080

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0080
Availability details for
Quantity
  • Availability
  • Add to Cart