COM448685539 Combi-Blocks, Inc.

3-(3,5-Difluorophenyl)-5-trifluoromethylbenzoic acid

  • MDL number MFCD18320000
  • Cas Number : 1261950-34-8
  • Molecular Weight : 302.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0160

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0160
Availability details for
Quantity
  • Availability
  • Add to Cart