COM448685542 Combi-Blocks, Inc.

4-(2,5-Difluorophenyl)-2-fluorobenzoic acid

  • MDL number MFCD12859419
  • Cas Number : 1184513-36-7
  • Molecular Weight : 252.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0235

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0235
Availability details for
Quantity
  • Availability
  • Add to Cart