COM448685543 Combi-Blocks, Inc.

3-(3-Acetylphenyl)-4-fluorobenzoic acid

  • MDL number MFCD18320097
  • Cas Number : 1261991-51-8
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0264

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0264
Availability details for
Quantity
  • Availability
  • Add to Cart