COM448685545 Combi-Blocks, Inc.

4-(3-Acetylphenyl)-2-fluorobenzoic acid

  • MDL number MFCD18320099
  • Cas Number : 1261906-36-8
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0266
Availability details for
Quantity
  • Availability
  • Add to Cart