COM448685551 Combi-Blocks, Inc.

4-(3-Carboxyphenyl)-3-methylbenzoic acid

  • MDL number MFCD18320242
  • Cas Number : 1261910-15-9
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0410

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0410
Availability details for
Quantity
  • Availability
  • Add to Cart