COM448685556 Combi-Blocks, Inc.

4-(4-Carboxyphenyl)-2-hydroxybenzoic acid

  • MDL number MFCD18320275
  • Cas Number : 861600-11-5
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0447

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0447
Availability details for
Quantity
  • Availability
  • Add to Cart