COM448685560 Combi-Blocks, Inc.

4-(4-Carboxyphenyl)-2-chlorobenzoic acid

  • MDL number MFCD18320286
  • Cas Number : 1261891-38-6
  • Molecular Weight : 276.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 97

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0462
Availability details for
Quantity
  • Availability
  • Add to Cart