COM448685561 Combi-Blocks, Inc.

3-(4-Carboxyphenyl)-5-nitrobenzoic acid

  • MDL number MFCD18320292
  • Cas Number : 1261931-45-6
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0468

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0468
Availability details for
Quantity
  • Availability
  • Add to Cart