COM448685563 Combi-Blocks, Inc.

3-Amino-5-(3,4-methylenedioxyphenyl)benzoic acid

  • MDL number MFCD18320298
  • Cas Number : 1261906-95-9
  • Molecular Weight : 257.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0475

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0475
Availability details for
Quantity
  • Availability
  • Add to Cart