COM448685564 Combi-Blocks, Inc.

5-Hydroxy-3-(3,4-methylenedioxyphenyl)benzoic acid

  • MDL number MFCD18086730
  • Cas Number : 1258628-34-0
  • Molecular Weight : 258.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0477

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0477
Availability details for
Quantity
  • Availability
  • Add to Cart