COM448685565 Combi-Blocks, Inc.

5-Methoxy-3-(3,4-methylenedioxyphenyl)benzoic acid

  • MDL number MFCD18320310
  • Cas Number : 1261960-47-7
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0490

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0490
Availability details for
Quantity
  • Availability
  • Add to Cart