COM448685566 Combi-Blocks, Inc.

4-Chloro-3-(3,4-methylenedioxyphenyl)benzoic acid

  • MDL number MFCD11933784
  • Cas Number : 1181596-03-1
  • Molecular Weight : 276.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0497

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0497
Availability details for
Quantity
  • Availability
  • Add to Cart