COM448685567 Combi-Blocks, Inc.

3-(3,4-Methylenedioxyphenyl)-5-nitrobenzoic acid

  • MDL number MFCD18320318
  • Cas Number : 1261894-05-6
  • Molecular Weight : 288.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0499
Availability details for
Quantity
  • Availability
  • Add to Cart