COM448685569 Combi-Blocks, Inc.

3-(4-Methoxy-2-methylphenyl)-2-methylbenzoic acid

  • MDL number MFCD18320348
  • Cas Number : 1261899-37-9
  • Molecular Weight : 256.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0535

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0535
Availability details for
Quantity
  • Availability
  • Add to Cart