COM448685573 Combi-Blocks, Inc.

2-Chloro-4-(3-nitrophenyl)benzoic acid

  • MDL number MFCD18320443
  • Cas Number : 1261894-36-3
  • Molecular Weight : 277.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0648

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0648
Availability details for
Quantity
  • Availability
  • Add to Cart