COM448685574 Combi-Blocks, Inc.

3-Methyl-4-(2-methylthiophenyl)benzoic acid

  • MDL number MFCD18320453
  • Cas Number : 1192547-93-5
  • Molecular Weight : 258.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0658

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0658
Availability details for
Quantity
  • Availability
  • Add to Cart