COM448685581 Combi-Blocks, Inc.

3-(4-Methylthiophenyl)-5-nitrobenzoic acid

  • MDL number MFCD18320542
  • Cas Number : 1261966-64-6
  • Molecular Weight : 290.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0747

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0747
Availability details for
Quantity
  • Availability
  • Add to Cart