COM448685582 Combi-Blocks, Inc.

3-(4-Methylthiophenyl)-5-trifluoromethylbenzoic acid

  • MDL number MFCD18320544
  • Cas Number : 1261975-51-2
  • Molecular Weight : 312.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0749

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0749
Availability details for
Quantity
  • Availability
  • Add to Cart