COM448685584 Combi-Blocks, Inc.

2-Chloro-4-(4-fluoro-3-methoxyphenyl)benzoic acid

  • MDL number MFCD18320679
  • Cas Number : 1261892-14-1
  • Molecular Weight : 280.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0896

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0896
Availability details for
Quantity
  • Availability
  • Add to Cart