COM448685587 Combi-Blocks, Inc.

4-(2-Chloro-4-methylphenyl)-3-fluorobenzoic acid

  • MDL number MFCD18320720
  • Cas Number : 1262008-90-1
  • Molecular Weight : 264.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0947

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0947
Availability details for
Quantity
  • Availability
  • Add to Cart