COM448685588 Combi-Blocks, Inc.

3-(2-Chloro-4-methylphenyl)-5-nitrobenzoic acid

  • MDL number MFCD18320737
  • Cas Number : 1261967-52-5
  • Molecular Weight : 292.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0964

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-0964
Availability details for
Quantity
  • Availability
  • Add to Cart