COM448685593 Combi-Blocks, Inc.

4-(3-Chloro-5-fluorophenyl)-3-fluorobenzoic acid

  • MDL number MFCD18320999
  • Cas Number : 1261970-29-9
  • Molecular Weight : 268.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1226

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1226
Availability details for
Quantity
  • Availability
  • Add to Cart