COM448685596 Combi-Blocks, Inc.

4-(3-Chloro-4-fluorophenyl)-3-fluorobenzoic acid

  • MDL number MFCD18321030
  • Cas Number : 1261922-99-9
  • Molecular Weight : 268.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1257

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1257
Availability details for
Quantity
  • Availability
  • Add to Cart