COM448685597 Combi-Blocks, Inc.

3-Methyl-4-(4-t-butylphenyl)benzoic acid

  • MDL number MFCD18321107
  • Cas Number : 1261923-72-1
  • Molecular Weight : 268.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1340

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1340
Availability details for
Quantity
  • Availability
  • Add to Cart