COM448685598 Combi-Blocks, Inc.

3-Fluoro-4-(4-t-butylphenyl)benzoic acid

  • MDL number MFCD18321113
  • Cas Number : 1261954-92-0
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1350

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1350
Availability details for
Quantity
  • Availability
  • Add to Cart