COM448685600 Combi-Blocks, Inc.

2-Chloro-5-(4-t-butylphenyl)benzoic acid

  • MDL number MFCD09042265
  • Cas Number : 926201-32-3
  • Molecular Weight : 288.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 97

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1360
Availability details for
Quantity
  • Availability
  • Add to Cart