COM448685604 Combi-Blocks, Inc.

2-Fluoro-5-(3-methoxycarbonylphenyl)benzoic acid

  • MDL number MFCD18321202
  • Cas Number : 1261912-57-5
  • Molecular Weight : 274.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1445

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1445
Availability details for
Quantity
  • Availability
  • Add to Cart