COM448685605 Combi-Blocks, Inc.

2-Chloro-5-(3-methoxycarbonylphenyl)benzoic acid

  • MDL number MFCD18321210
  • Cas Number : 1262009-62-0
  • Molecular Weight : 290.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1453

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1453
Availability details for
Quantity
  • Availability
  • Add to Cart