COM448685606 Combi-Blocks, Inc.

2-Chloro-4-(3-methoxycarbonylphenyl)benzoic acid

  • MDL number MFCD18321211
  • Cas Number : 1262004-76-1
  • Molecular Weight : 290.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1454

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1454
Availability details for
Quantity
  • Availability
  • Add to Cart