COM448685608 Combi-Blocks, Inc.

2-Fluoro-5-(4-methoxycarbonylphenyl)benzoic acid

  • MDL number MFCD18321232
  • Cas Number : 1261980-30-6
  • Molecular Weight : 274.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1476

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1476
Availability details for
Quantity
  • Availability
  • Add to Cart