COM448685609 Combi-Blocks, Inc.

2-Chloro-5-(4-methoxycarbonylphenyl)benzoic acid

  • MDL number MFCD18321240
  • Cas Number : 1261903-35-8
  • Molecular Weight : 290.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1484

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1484
Availability details for
Quantity
  • Availability
  • Add to Cart