COM448685611 Combi-Blocks, Inc.

3-(2,4-Dimethoxyphenyl)-2-methylbenzoic acid

  • MDL number MFCD18321314
  • Cas Number : 1261912-87-1
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1558

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1558
Availability details for
Quantity
  • Availability
  • Add to Cart