COM448685612 Combi-Blocks, Inc.

4-(2,4-Dimethoxyphenyl)-2-fluorobenzoic acid

  • MDL number MFCD18321323
  • Cas Number : 1179916-77-8
  • Molecular Weight : 276.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1568

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1568
Availability details for
Quantity
  • Availability
  • Add to Cart