COM448685616 Combi-Blocks, Inc.

4-(3,5-Dimethoxyphenyl)-2-fluorobenzoic acid

  • MDL number MFCD18321409
  • Cas Number : 1261980-91-9
  • Molecular Weight : 276.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1661

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1661
Availability details for
Quantity
  • Availability
  • Add to Cart