COM448685618 Combi-Blocks, Inc.

4-(3-Carboxy-5-fluorophenyl)-2-fluorobenzoic acid

  • MDL number MFCD18321471
  • Cas Number : 1261978-52-2
  • Molecular Weight : 278.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1723

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1723
Availability details for
Quantity
  • Availability
  • Add to Cart