COM448685619 Combi-Blocks, Inc.

4-(3-Carboxy-5-fluorophenyl)-2-chlorobenzoic acid

  • MDL number MFCD18321481
  • Cas Number : 1261906-07-3
  • Molecular Weight : 294.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1733
Availability details for
Quantity
  • Availability
  • Add to Cart