COM448685623 Combi-Blocks, Inc.

4-(4-Carboxy-3-fluorophenyl)-2-fluorobenzoic acid

  • MDL number MFCD18321501
  • Cas Number : 1261929-07-0
  • Molecular Weight : 278.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1754

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1754
Availability details for
Quantity
  • Availability
  • Add to Cart