COM448685625 Combi-Blocks, Inc.

3-(4-Carboxy-3-fluorophenyl)-5-methoxybenzoic acid

  • MDL number MFCD18321507
  • Cas Number : 1261970-06-2
  • Molecular Weight : 290.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1761

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1761
Availability details for
Quantity
  • Availability
  • Add to Cart