COM448685626 Combi-Blocks, Inc.

4-(4-Carboxy-3-fluorophenyl)-2-chlorobenzoic acid

  • MDL number MFCD18321510
  • Cas Number : 1261964-19-5
  • Molecular Weight : 294.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1764

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1764
Availability details for
Quantity
  • Availability
  • Add to Cart