COM448685627 Combi-Blocks, Inc.

3-(4-Carboxy-3-fluorophenyl)-5-nitrobenzoic acid

  • MDL number MFCD18321516
  • Cas Number : 1261970-11-9
  • Molecular Weight : 306.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1770

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1770
Availability details for
Quantity
  • Availability
  • Add to Cart