COM448685633 Combi-Blocks, Inc.

4-(2-Chloro-5-methoxyphenyl)-3-fluorobenzoic acid

  • MDL number MFCD18321620
  • Cas Number : 1261904-84-0
  • Molecular Weight : 280.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1877

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1877
Availability details for
Quantity
  • Availability
  • Add to Cart