COM448685634 Combi-Blocks, Inc.

3-(2-Chloro-5-methoxyphenyl)-5-nitrobenzoic acid

  • MDL number MFCD18321637
  • Cas Number : 1261892-79-8
  • Molecular Weight : 308.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1894

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1894
Availability details for
Quantity
  • Availability
  • Add to Cart