COM448685636 Combi-Blocks, Inc.

2-(5-Chloro-2-methoxyphenyl)-5-fluorobenzoic acid

  • MDL number MFCD12859231
  • Cas Number : 1178334-17-2
  • Molecular Weight : 280.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1968

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-1968
Availability details for
Quantity
  • Availability
  • Add to Cart