COM448685643 Combi-Blocks, Inc.

4-Fluoro-3-(3-trifluoromethylphenyl)benzoic acid

  • MDL number MFCD18321760
  • Cas Number : 1261593-39-8
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2031

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2031
Availability details for
Quantity
  • Availability
  • Add to Cart