COM448685646 Combi-Blocks, Inc.

2-Fluoro-5-(3-trifluoromethylphenyl)benzoic acid

  • MDL number MFCD09042263
  • Cas Number : 926235-51-0
  • Molecular Weight : 284.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2034

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2034
Availability details for
Quantity
  • Availability
  • Add to Cart