COM448685656 Combi-Blocks, Inc.

5-Chloro-3-(2,4-dichlorophenyl)benzoic acid

  • MDL number MFCD18321851
  • Cas Number : 1261911-18-5
  • Molecular Weight : 301.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2137

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2137
Availability details for
Quantity
  • Availability
  • Add to Cart