COM448685659 Combi-Blocks, Inc.

3-(3,4-Dichlorophenyl)-5-trifluoromethylbenzoic acid

  • MDL number MFCD18321903
  • Cas Number : 1261930-84-0
  • Molecular Weight : 335.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2206

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2206
Availability details for
Quantity
  • Availability
  • Add to Cart