COM448685660 Combi-Blocks, Inc.

4-(3,5-Dichlorophenyl)-2-fluorobenzoic acid

  • MDL number MFCD18321916
  • Cas Number : 1261913-97-6
  • Molecular Weight : 285.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2219

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YB-2219
Availability details for
Quantity
  • Availability
  • Add to Cart